Sodium hydroxide 50% w/v
Supplier: Ricca Chemical
|
Danger
|
Synonyms:
Caustic soda
R7282000-20F
R7282000-4A
RCR7282000-20FEA
2422.14
CAD
RCR7282000-20F
RCR7282000-4A
Sodium hydroxide 50% w/v
Sodium hydroxide
Formula:
NaOH MW: 40 g/mol Boiling Pt: 1390 Melting Pt: 323 Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1824 ADR: 8 II Merck Index: 14,08627 |
Specification Test Results
Appearance | Colorless liquid |
Assay (vs. Sulfuric Acid/Phenolphthalein) | 49.92 - 50.08 % (w/v) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...