Sodium hydroxide 10% (w/w)

Supplier: Ricca Chemical
Danger

R7261000-1A R7261000-20F R7261000-4A
RCR7261000-1AEA 77.42 CAD
RCR7261000-1A RCR7261000-20F RCR7261000-4A
Sodium hydroxide 10% (w/w)
Sodium hydroxide
Formula: NaOH
Boiling Pt: ~100 °C
Melting Pt: ~0 °C
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Sulfuric Acid/Phenolphthalein) 9.9 - 10.1 % (w/w)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR