Axygen® Deep Well and Assay Plates, Corning
Supplier: Corning
P-DW-20-C-S P-5ML-48-C P-5ML-48-C-S P-DW-11-C-S P-384-240SQ-C P-96-450R-C P-96-450V-C-S P-384-120SQ-C-S P-384-240SQ-C-S P-2ML-SQ-C-S P-DW-20-C-S-IND P-DW-11-C P-DW-20-C P-384-120SQ-C P-96-450R-C-S P-96-450V-C P-2ML-SQ-C P-DW-500-C P-DW-12-HC P-DW-12-HC-S
10011-944CS 685.17 CAD
10011-944 12000-726 12000-728 47734-788 47743-466 47743-982 47743-998 47745-400 47745-404 47749-874 47749-930 89182-904 10011-940 10011-942 47743-468 47743-984 47743-996 47744-806 89182-902 89230-174 89230-176
Axygen® Deep Well and Assay Plates, Corning
Microplates Deep Well Microplates
Axygen® deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage.
  • 0.6 ml deep well plate
  • ANSI/SLAS 1-2004: Microplates - footprint dimensions
  • Ultra-flat surface ensures proper sealing with heat sealing film
Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to −80 °C.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification I-RECs, ISO 14001 - Reynosa