Corning® Standard Polystyrene Microplates, 1536-well
Supplier: Corning
3891 3893 7246 7247 7248 7249 3937 3936 3728 3724 3726 3895 3729 3725 3727 3838BC 3731BC 3831 3838 3832BC 3833 3835 3836 3839BC
66025-652CS 2703.07 CAD
66025-652 66025-654 71001-346 71001-348 71001-350 71001-352 77776-916 77776-918 89089-724 89089-728 89089-730 89089-732 89089-738 89089-742 89089-746 89126-916 89126-918 89130-726 89130-730 89130-734 89218-978 89218-980 89218-982 30621-094
Corning® Standard Polystyrene Microplates, 1536-well
Microplates HTS Microplates
These PS microplates conform to standard microplate footprint and dimensions, and are available in solid black or white, with round or flat bottoms, or in black clear bottom formats.
  • Total well volume of 10 μl for round well plates and 12.8 μl for flat bottom plates
  • Recommended working volume of up to 8 μl
  • Round well bottoms for reduced air entrapment and improved CVs and Z factor
  • Raised well bottoms for higher sensitivity
  • Lids available separately
* NBS™ = Non binding surface