Corning® 96-well Clear Polystyrene Microplates, Corning
Supplier: Corning
3358 3366 3369 3370 3585 3598 3599 3696 3894 3590 3591 3690 9018 3795 3797 3798 3896 3897 3898 3474 3997 3628 3697 3695 3641 3788 3300 3628BC 7007 3841 3368 3595 3596 3799 9017 3367 3360 3361 3797 9018BC
25381-048CS 839.95 CAD
25381-048 25381-050 25381-054 25381-056 29442-050 29442-058 29442-060 29442-062 29442-068 29442-314 29442-316 29442-318 29442-322 29442-392 29442-396 29442-398 29442-400 29442-402 29442-404 29443-034 29443-954 29444-274 29444-316 29444-320 29445-142 29445-154 66025-626 71001-358 89089-826 89092-706 25381-052 29442-054 29442-056 29442-066 29442-070 29444-288 29444-454 29444-456 470211-270 89501-318
Corning® 96-well Clear Polystyrene Microplates, Corning
Microplates Cell Culture Plates
Cell culture microplates are sterile and nonpyrogenic. Lids available where indicated.
NA Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays. NA Cat. Nos. 3585 and 3595 have special low evaporation lids. NA Cat. No. 3841 is aseptically manufactured.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification RECs, ISO 14001 - Kennebunk