Corning® BioCoat® Poly-D-Lysine Culture Dishes, Corning
Supplier: Corning
356467 356468 356469 354467 354468 354469 354550
CACB356467ACS 562.7 CAD
CACB356467A CACB356468A CACB356469A CA-CB354467 CA-CB354468 CA-CB354469 CA-CB354550
Corning® BioCoat® Poly-D-Lysine Culture Dishes, Corning
Culture Dishes Cell Culture Dishes
Poly-D-Lysine (PDL) and Poly-L-Lysine (PLL) are synthetic compounds that enhance cell adhesion and protein absorption by altering surface charges on the culture substrate. In addition to promoting cell adhesion, poly-lysine surface treatments support neurite outgrowth and improve the survival of many central nervous system (CNS) primary cells in culture. As PDL and PLL are synthetic molecules, they do not stimulate biological activity in the cells cultured on them, and they do not introduce impurities carried by natural polymers.
  • Attachment and spreading of a variety of cell types
  • Cell
More Product Information