Corning® BioCoat® Poly-L-Lysine-coated Culture Dishes, Corning
Supplier: Corning
356517 354517 356518 354518
CA62406-292CS 479.21 CAD
CA62406-292 CA62406-268 CA62406-270 CA62406-272
Corning® BioCoat® Poly-L-Lysine-coated Culture Dishes, Corning
Culture Dishes Cell Culture Dishes
Poly-D-Lysine (PDL) and Poly-L-Lysine (PLL) are synthetic compounds that enhance cell adhesion and protein absorption by altering surface charges on the culture substrate. In addition to promoting cell adhesion, poly-lysine surface treatments support neurite outgrowth and improve the survival of many central nervous system (CNS) primary cells in culture. As PDL and PLL are synthetic molecules, they do not stimulate biological activity in the cells cultured on them, and they do not introduce impurities carried by natural polymers.
  • Attachment and spreading of a variety of cell types
  • Cell
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Environmental certification