Corning® BioCoat® Poly-D-Lysine Culture Slides, Corning
Supplier: Corning
354577 354632
CACB354577CS 538.12 CAD
CACB354577 CACB354632
Corning® BioCoat® Poly-D-Lysine Culture Slides, Corning
Microplates Cell Culture Plates
Poly-D-Lysine (PDL) and Poly-L-Lysine (PLL) are synthetic compounds that enhance cell adhesion and protein absorption by altering surface charges on the culture substrate. In addition to promoting cell adhesion, poly-lysine surface treatments support neurite outgrowth and improve the survival of many central nervous system (CNS) primary cells in culture. As PDL and PLL are synthetic molecules, they do not stimulate biological activity in the cells cultured on them, and they do not introduce impurities carried by natural polymers.
  • Attachment and spreading of a variety of cell types
  • Cell
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Environmental certification