Corning® BioCoat® Collagen I-coated Culture Slides, Corning

Supplier: Corning
354557 354630
CACB354557CS 680.14 CAD
CACB354557 CACB354630
Corning® BioCoat® Collagen I-coated Culture Slides, Corning
Microplates Cell Culture Plates
Collagen I, found in most tissues and organs, is most plentiful in dermis, tendon, and bone. It is an integral part of the framework that holds cells and tissues together and has been recognized as a useful matrix for improving cell culture. in vitro use of collagen can exert effects on the adherence, morphology, growth, migration, and differentiation of a variety of cell types.

  • Promotion of cell attachment and spreading
  • Rapid expansion of cell populations
  • Serum-free or reduced serum culture
  • Cell adhesion assays
  • Improving survival of primary cells in culture
  • Source: Rat tail tendon
  • Storage: 4 to 30 °C under dry conditions.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR