Corning® BioCoat® Collagen I-coated Culture Dishes, Corning
Supplier: Corning
356401 356450 356456 354450 354456 354551 354401
CACB356401CS 596.22 CAD
CACB356401 CACB356450 CACB356456 CA-CB354450 CA-CB354456 CA-CB354551 CACB354401
Corning® BioCoat® Collagen I-coated Culture Dishes, Corning
Culture Dishes Cell Culture Dishes
Collagen I, found in most tissues and organs, is most plentiful in dermis, tendon, and bone. It is an integral part of the framework that holds cells and tissues together and has been recognized as a useful matrix for improving cell culture. in vitro use of collagen can exert effects on the adherence, morphology, growth, migration, and differentiation of a variety of cell types.
  • Promotion of cell attachment and spreading
  • Rapid expansion of cell populations
  • Serum-free or reduced serum culture
  • Cell adhesion assays
  • Improving survival of primary cells in
More Product Information