VWR® Tube Revolvers
Supplier: VWR International
76595-822 76595-824
76595-822EA 295.86 CAD
76595-822 76595-824
VWR® Tube Revolvers
Shakers Rotating/Rocking Shakers
Efficient, powerful tube revolvers fixed or variable speed, with 5 interchangeable rotor series.
  • Choose from fixed speed, 25 rpm or variable speed, 10 to 40 rpm models
  • Variable speed model with LED display can be programmed to oscillate at every 40⁰ to simulate hand mixing with rotation and shaking
  • Includes 5 interchangeable rotisseries to accommodate a variety of tube sizes simultaneously, each paddle can be adjusted from 0 to 90⁰
  • Ergonomically designed for simple operation and minimal bench space usage
  • Maintenance free motor is powerful and quiet
  • 2-year
More Product Information