VWR® Tube Rotators
Supplier: VWR International
76595-826 76595-828 76595-830
76595-826EA 769.25 CAD
76595-826 76595-828 76595-830
VWR® Tube Rotators
Shakers Rotating/Rocking Shakers
VWR® Tube rotators are ideal for applications requiring highly efficient and consistent mixing. VWR offers rugged and reliable fixed speed, digital and large capacity models.
  • Adjustable tilt angle to accommodate different applications
  • Variety of VWR® tube holders, carousels and drums available to accommodate various size sample tubes efficiently
  • All units are suitable for use in cold room and incubator environments
  • Digital tube rotator and the large capacity tube rotator offer adjustable speed, continuous and timer modes, with automatic stop
  • 2 Drums available for 14 or 18 mm culture tubes for the large capacity tube rotator
  • 2 year replacement warranty
Ideally suited for clinical, research, and academic applications.
Certifications: CE certified
Delivery information: Units 76595-826 and 76595-826 supplied with VWR® carousel for 12×50 ml tubes (76595-856), large capacity model 76595-830 is suplied with carousel for 15x50 ml tubes (76595-862) plus instruction manual and power cord.