Corning® 96-Well Solid Black and White Polystyrene Microplates, Corning
Supplier: Corning
3605 3792 3912 3922 3925 3916 3917 3688 3693 3694 3362 3650 3642 3991 3993 3875 3686 3600 3915 3990 4591 3789A
16001-310CS 1604.61 CAD
16001-310 29442-922 29444-016 29444-020 29444-022 29444-039 29444-041 29444-308 29444-312 29444-314 29444-458 29445-140 33501-808 33501-812 33501-814 33501-816 33501-834 77776-924 89091-016 29444-018 33501-810 10037-562 75874-852
Corning® 96-Well Solid Black and White Polystyrene Microplates, Corning
Microplates Cell Culture Plates
These 96-well solid black and white polystyrene microplates are designed to reduce well-to-well cross-talk.
White microplates enhance luminescent signals and have low background luminescence.
Black microplates have low background fluorescence and minimize light scattering.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification RECs, ISO 14001 - Kennebunk