Corning® BioCoat® Gelatin Microplates, Corning
Supplier: Corning
354689 356689
CABD354689CS 160.86 CAD
CABD354689 CABD356689
Corning® BioCoat® Gelatin Microplates, Corning
Microplates Cell Culture Plates
PS coated with porcine gelatin. Corning® BioCoat™ gelatin provides an attachment and growth promoting substrate for the culture of a variety of cell types. Gelatin is commonly used in the culture of normal and transfected cell types, including vascular endothelial, muscle, embryonic stem (ES) and F9 teratocarcinoma cells.
  • Gelatin substrate enhances the attachment of a variety of normal and transfected cell types
  • Pre-treatment with gelatin eliminates time consuming preparation, saving time and money
  • Lot-to-lot consistency ensures reproducible results
Order Now
SPECIFICATIONS
Environmentally Preferable
Environmental certification