Transwell® Clear Inserts, Polyester (PET) Membrane
Supplier: Corning
3450 3452 3460 3470 3472 3462 3464
29442-074CS 408.03 CAD
29442-074 29442-076 29442-078 29442-082 29442-084 29442-080 89235-020
Transwell® Clear Inserts, Polyester (PET) Membrane
Microplates Insert Plates and Inserts
Permeable supports, also known as cell culture inserts, are an essential tool for the study of both anchorage-dependent and independent cell lines.
  • w Produce a cell culture environment that closely resembles an in vivo state
  • Co-culture cells with or without cell-to-cell contact
  • Design a diversity of experiments using various pore sizes, membrane types, and coatings 10 µm transparent membrane
  • Treated for optimal cell attachment
  • Excellent visibility under phase contrast microscopy
  • Supplied in multiple well plates
  • Sterile
Allow polarized cells to carryMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification RECs, ISO 14001 - Kennebunk