Corning® BioCoat® Poly-D-Lysine Microplates, Corning
Supplier: Corning
356691 356620 356663 356662 356661 356660 356651 354663 354662 354651 354461 354640 354620 354660 354661 356690 356697 356461 356640
CA47748-264CS 1535.59 CAD
CA47748-264 CA62406-282 CA62406-350 CA62406-354 CA62406-358 CA62406-362 CA62406-366 CA-62406-352 CA-62406-356 CA-62406-368 CA-CB354461 CA-CB354640 CA62406-284 CA62406-364 CA62406-361 CA47748-262 CA47748-266 CA47748-276 CA62406-032 CA62406-036
Corning® BioCoat® Poly-D-Lysine Microplates, Corning
Microplates Cell Culture Plates
Poly-D-Lysine (PDL) and Poly-L-Lysine (PLL) are synthetic compounds that enhance cell adhesion and protein absorption by altering surface charges on the culture substrate. In addition to promoting cell adhesion, poly-lysine surface treatments support neurite outgrowth and improve the survival of many central nervous system (CNS) primary cells in culture. As PDL and PLL are synthetic molecules, they do not stimulate biological activity in the cells cultured on them, and they do not introduce impurities carried by natural polymers.
  • Attachment and spreading of a variety of cell types
  • Cell differentiation and neurite outgrowth
  • Attachment of fastidious transfected cell lines
  • Support survival of primary neurons in culture
  • Serum-free or reduced-serum culture
  • Storage: 4 to 30 °C under dry conditions