Corning® BioCoat® Poly-D-Lysine Flasks, Corning
Supplier: Corning
356536 356537 356538 356539 354538 354536 354537 354539
CA12777-090CS 813.05 CAD
CA12777-090 CA12777-094 CA12777-096 CA12777-100 CA-12777-084 CA-12777-088 CA-12777-092 CA-12777-098
Corning® BioCoat® Poly-D-Lysine Flasks, Corning
Flasks Cell Culture Flasks
Poly-D-Lysine (PDL) and Poly-L-Lysine (PLL) are synthetic compounds that enhance cell adhesion and protein absorption by altering surface charges on the culture substrate. In addition to promoting cell adhesion, poly-lysine surface treatments support neurite outgrowth and improve the survival of many central nervous system (CNS) primary cells in culture. As PDL and PLL are synthetic molecules, they do not stimulate biological activity in the cells cultured on them, and they do not introduce impurities carried by natural polymers.
  • Attachment and spreading of a variety of cell types
  • Cell
More Product Information