Falcon® Bulk Packaged Serological Pipettes, Corning
Supplier: Corning
357506 357508 357530 357515
CA53300-261CS 647.14 CAD
CA53300-261 CA53300-365 CA53300-501 CA53392-198
Falcon® Bulk Packaged Serological Pipettes, Corning
Pipettes Serological Pipettes
The pipette graduation alignment system is a fast and simple way to bring graduations to your line of sight. Polystyrene, disposable pipettes for tissue culture, bacteriological, and research applications.
  • Sterile
  • RNase-/DNase-free
  • Noncytotoxic, nonpyrogenicity tested to less than 0.1 EU/ml
  • Color-coded package and markings for ease of identification
Simply pop open or peel back the top of the pipette package and line up the pipettor before completely removing the pipette from its wrapping. When you remove the pipette from its package, the graduationsMore Product Information