PureAmp PCR and ELISA Plate Sealing Films
Supplier: MTC BIO, INC.
P1001-A P1001-E P1001-PCR P1001-Q P1001-QPS
470347-462PK 145.05 CAD
470347-462 470347-468 470347-466 470347-464 470347-470
PureAmp PCR and ELISA Plate Sealing Films
Seals Microplate Seals
A versatile sealing film for standard ELISA and PCR 96-well plates.
  • RNase, DNase, DNA, and PCR inhibitor free
  • EZ-Peel adhesive for spill-free removal
  • Inhibits evaporation across all 96 wells (≤5%)
  • Suitable for a wide temperature range
The sealing options in the PureAmp™ line include clear plastic film for end-point PCR, two optically clear films for qPCR, plus one pierceable aluminum membrane for storage and/or PCR. All PureAmp™ sealing products are pre-cut for a perfect fit on the plate every time and are guaranteed to keep evaporation rates under 5%More Product Information