Corning® Microcarriers for Bioprocess Scale-up, Corning
Supplier: Corning
3781 3779 3772 3786 4621 4622 4623 4624 4620 4625
30617-552CS 469.58 CAD
30617-552 30617-554 89499-404 89499-408 10002-704 10002-706 10002-708 10002-710 10002-712 10002-714
Corning® Microcarriers for Bioprocess Scale-up, Corning
Beads Microcarriers
Corning® Microcarriers are designed to simplify bioprocessing scale up. The microcarriers are sterile, ready to use, and available in closed systems packaging to be used directly with bioreactors. Additionally, the microcarriers are surface treated to enhance cell attachment, maximising cell yield and viability.
  • USP Class VI polystyrene material provides a consistent platform
  • Available with a choice of surface treatments
  • Sterile (SAL 10⁻⁶)
  • Certified non pyrogenic
  • Available as closed system solutions
Corning® Microcarriers are available with a choice of surface treatments. Corning Synthemax® II substrate creates a synthetic surface on the microcarriers for stem cell expansion. It is recommended to use Low Concentration for MSC applications and High Concentration for iPS/ES applications. Corning CellBIND® surface treatment infuses the surface of the microcarriers with oxygen to improve cell attachment. Corning® Microcarriers with a positive charge surface treatment or collagen coating are also available.
Bead size: 125 to 212 µm
Density: 1.026 ±0.0004 g/cm³
Surface area: 360 cm² /g