Eisco Glass Burettes with PTFE Key Stopcock
Supplier: Wards
CH0241L CH0240A CH0240D CH0240G CH0240H CH0240F CH0241K CH0240B CH0241J CH0240C
470343-688EA 70 CAD
470343-688 470343-832 470343-934 470344-068 470344-472 470344-508 470344-576 470345-046 470345-796 470346-096
Eisco Glass Burettes with PTFE Key Stopcock
Burettes
PTFE key stopcock offers superior chemical resistance and low therma.
  • Class A and Class B versions available
  • Made from high quality 3.3 borosilicate glass
  • Straight single bore
  • Blue enamel graduations for contrast against clear liquids
These burettes are made of precisely-bore, high quality, heavy duty borosilicate 3.3 glass with excellent durability and chemical resistance. Both Class A and Class B versions conform with DIN ISO 385 standards for accuracy. The attached PTFE key stopcock has high chemical, moisture, heat and UV resistance. Calibrated to deliver.