Eisco Premium Glass Burettes with Glass Key Stopcock
Supplier: Wards
CH0236F CH0236E CH0236H CH0236C CH0236G CH0236D CH0236A CH0236B
470343-656EA 30.8 CAD
470343-656 470344-026 470344-028 470344-308 470344-344 470344-558 470344-608 470344-880
Eisco Premium Glass Burettes with Glass Key Stopcock
Burettes
All-glass stopcock for smooth motion and flow adjustment.
  • Class A and Class B versions available
  • Made from high quality 3.3 borosilicate glass
  • Blue enamel graduations for contrast against clear liquids
  • Straight single bore
These premium burettes are made of precisely bore high quality, heavy duty borosilicate 3.3 glass with excellent durability and chemical resistance. Both Class A and Class B versions conform with DIN ISO 385 standards for accuracy. Each burette includes an attached key stopcock made of high quality glass and features blue graduations and markings. To avoid material damaging friction between the burette and the stopcock and to ensure tightness joint, grease is necessary when working with burettes with glass stopcock. Calibrated to deliver.