Eisco Automatic Zero Glass Burettes
Supplier: Wards
CH0242E CH0242C CH0242D CH0242A CH0242G CH0242H CH0242B CH0242F
470343-634EA 253.1 CAD
470343-634 470343-674 470343-952 470344-104 470344-416 470344-430 470344-930 470345-920
Eisco Automatic Zero Glass Burettes
Burettes
Custom foam packaging for safe transport and storage.
  • Automatic zero burette mounted on a glass bottle reservoir via ground glass joint
  • Designed to automatically set to zero mark upon pressure release
  • Both burette and reservoir are made of heavy-duty borosilicate 3.3 glass
  • Class A and Class B versions available
  • Includes PTFE key stopcock
  • White enamel graduations for contrast against dark liquids
Automatic Zero Class A burettes are mounted on a graduated reservoir bottle. Solution rises to the top of burette by squeezing the rubber bulb. The burette is automatically set to zero releasing the air pressure from the valve. Both burette and reservoir are made of precisely-bore, high quality, heavy-duty borosilicate 3.3 glass with excellent durability and chemical resistance. Both Class A and Class B versions conform with DIN ISO 385 standards for accuracy. Calibrated to deliver. All components are packed in custom foam inserts for safe transport and storage.