CiDehol® 70 Pre-Saturated Cleanroom Wipes, Decon Labs
Supplier: Decon Labs
8776
76579-806CS 663.09 CAD
76579-806
CiDehol® 70 Pre-Saturated Cleanroom Wipes, Decon Labs
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
CiDehol® 70 wipes are pre-saturated with 70% isopropyl alcohol.
  • Tear-resistant
  • Lint-free
  • Unique self-closing lid keeps wipes saturated
CiDehol® 70 wipes are strong, lint-free wipes made for use on hard surfaces in cleanrooms, labs, and other areas where a pre-saturated cleaning wipe is a convenience. Wipes are 100% PET, non-woven. Each wipe measures 5×8". Packaged 175 wipes per canister.