VWR® Rotary Evaporator Dry Ice Condenser
Supplier: VWR
VWR®
18900471
76595-784EA 3718.94 CAD
76595-784
VWR® Rotary Evaporator Dry Ice Condenser
Condensers
1000 ml rotary evaporator dry ice condenser, made of clear borosilicate glass.
  • Excellent chemical resistance, high temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
  • 1000 ml cooling surface
Delivery information: With evaporating flask (NS 24/40) and receiving flask (KS 35/20).