Masterflex® MasterSense® MP30 Reusable Pressure Transducers, Avantor®
Supplier: Avantor Fluid Handling
7526-80 7526-82
MFLX07526-80EA 1221.34 CAD
MFLX07526-80 MFLX07526-81 MFLX07526-82
Masterflex® MasterSense® MP30 Reusable Pressure Transducers, Avantor®
Pumps Peristaltic Pumps
High accuracy pressure transducers for plug and play connectivity with Masterflex® MasterSense® pump systems.
  • Add critical pressure monitoring to your workflow
  • All-stainless welded body and wetted parts
  • Rugged design for shock protection and excellent long-term stability
  • CE certificated
  • Sanitary models are designed for clean-in-place and sterilize-in-place
MasterSense® MP30 pressure transducers utilize a thin-film-on-steel measurement principle to provide an accurate 0 to 30 psi output directly to MasterSense® pump systems. Sensors include a six foot cable with AUX plug adaptor for easy plug and play.
Install downstream of pump to detect pressure changes in your process. Perfect for tangential-Flow filtration, depth filtration, harvest, bulk transfer, chromatography and fill applications. Choose the model with the appropriate process connection for your application.
Sensor MFLX07526-82 can be coupled with MFLX31203-33 single use tee and MFLX31201-95 single-use clamp for a gamma sterilizable flow path. Model MFLX07526-81 ³/₄" sanitary connection not recommended for applications with high temperature variations.
Certifications: CE Certified.