N,N-Dimethylformamide, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

BAKR9241-09 BAKR9241-37
CAJT9241-09EA 2886.9 CAD
CAJT9241-09 CAJT9241-37
N,N-Dimethylformamide, J.T.Baker®
N,N-Dimethylformamide
Formula: HCON(CH₃)₂
MDL Number: MFCD00003284
CAS Number: 68-12-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR