Evergreen 7" Chromatography Columns, Caplugs Evergreen

Supplier: Caplugs
208-3392-060 208-3393-060 208-3394-060
76498-832CT 383.5 CAD
76498-832 76498-834 76498-836
Evergreen 7" Chromatography Columns, Caplugs Evergreen
Chromatography Columns
Caplugs evergreen chromatography columns are ideal for use in general chromatographic work for extraction of components, with special applications in isoenzyme and protein studies, hemoglobin fractionation and desalting.

  • Precision-molded from premium-grade, chemically inert plastics
  • Polystyrene columns are glass-clear and appropriate for use with mild acidic (except oxidizing acids) and alkali solutions
  • Polypropylene columns are contact-transparent, more chemically resistant and appropriate for use with organic solvents and stronger acids (except oxidizing acids) and alkalis

Chromatography columns include reservoir and end caps and polyethylene filter matrix supports in one of three porosities:
- Fine (15 to 45 μm)
- Medium (45 to 90 μm)
- Coarse (90 to 135 μm)

Ordering information: Also available in 5" size.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR