Paraffin Wax Block Trimmer, Mopec
Supplier: Mopec
BK001
76579-208EA 3583.41 CAD
76579-208
Paraffin Wax Block Trimmer, Mopec
Paraffin Wax Trimmers and Dispensers
Mopec’s paraffin wax block trimmer is designed to easily remove excess paraffin from the edges and writing surfaces of embedding tissue cassettes without any damage to written text or barcodes.
  • Easily removes excess paraffin from tissue cassettes without any damage to written text or barcodes
  • Rapid heating system and extra large work surface allows multiple cassettes to be de-waxed simultaneously
  • Features digital temperature control with memory function for desired start-up temperature
  • Neutral slope accessible for both left and right-hand users
  • Dimensions: 6W×8L×7.25″ H
Users simply pass cassettes across the heated, grooved work station and wax is melted and collected in the disposable container at the bottom of the slope. The rapid heating system and extra large work surface allows multiple cassettes to be de-waxed simultaneously.