Biohazard Specimen Transport Bags, Mopec
Supplier: Mopec
BG682 BG682-P BG683 BG684
76579-398CS 272.71 CAD
76579-398 76579-400 76579-402 76579-404
Biohazard Specimen Transport Bags, Mopec
Bags Sample Bags
Mopec’s biohazard specimen transport bags are the perfect solution for safely and securely transporting various blood tubes, urine containers, or pathology specimens.
  • Used for transporting blood tubes, urine containers, or pathology specimens.
  • Ideal for use in hospitals and laboratories
  • Made from a high-quality LDPE material
  • Re-closable zipper
  • Separate pouch to safely store documents
  • Feature 'Biohazard' symbol
Ideal for use in hospitals and laboratories, these bags are made from a high-quality LDPE material, feature a re-closable zipper and have a separate pouch to safely store documents. Available in three sizes. 100 bags/pack. Sold by the case of 10 packs.