Eisco Amber Glass Dropping Bottles HDPE Stopper Head
Supplier: Wards
CH0195C CH0195A CH0195B
470343-922EA 25.9 CAD
470343-922 470344-518 470345-270
Eisco Amber Glass Dropping Bottles HDPE Stopper Head
Bottles Dropping Bottles
Highly resistant to acids and alkalis.
  • Made of high quality, heavy-duty, borosilicate 3.3 glass
  • 3-piece pipette section consists of a glass dropper, HDPE stopper with dust-proof head, and rubber bulb
  • Amber color glass appropriate for holding light sensitive contents
  • Suitable for a variety of applications in the classroom or laboratory
Equipped with an eye dropper pipette for clean dispensing of stored contents. The amber color glass absorbs UV waves, making it useful for light sensitive contents. Excellent for chemistry applications, transferring and mixing indicators, stains or other aqueous solutions.