Eisco Glass Reagent Bottles with Hexagonal Stoppers
Supplier: Wards
CH0162B CH0162F CH0162G CH0162A CH0162C CH0162E CH0162D
470344-322EA 13.09 CAD
470344-322 470344-584 470344-970 470345-668 470345-706 470345-808 470345-996
Eisco Glass Reagent Bottles with Hexagonal Stoppers
Bottles Glass Bottles
For holding chemicals in liquid or powder form.
  • Made of high-quality clear borosilicate 3.3 glass
  • Excellent durability and chemical resistance
  • Features a narrow neck to ensure a water and airtight seal
  • Includes a tight fitting, hexagonal hollow glass stopper
Autoclavable, with superior chemical durability. Low temperature gradient - can withstand high temperatures and thermal shocks.
Eisco borosilicate glass has the following heat tolerances:
- Strain Point 515 °C
- Annealing Point 565 °C
- Softening Point 820 °C
Order Now
SPECIFICATIONS
Environmentally Preferable