Plastic Graduated Beakers
Supplier: Wards
CH0137G CH0137AF CH0137BF CH0137CF CH0137CPK6 CH0137DF CH0137EF CH0137FF
470343-766EA 50.3 CAD
470343-766 470344-648 470344-868 470345-360 470345-418 470345-506 470345-700 470354-068 470354-070 470354-078 470354-076 470354-074 470354-066 470354-072
Plastic Graduated Beakers
Beakers
Autoclavable, microwavable, and dishwasher safe.
  • Made of premium quality polypropylene plastic
  • Superior chemical durability and heat tolerance
  • Excellent optical clarity
  • Raised 5 ml graduations and specifications
  • Tapered spout for easy pouring
Polypropylene plastic beakers offer excellent chemical durability. They are microwavable, autoclavable, and dishwasher safe for easy cleaning and sterilization; can withstand temperatures between −20 °C to 135 °C. Beakers have raised, five ml graduation lines. Pack of 12.
Order Now
SPECIFICATIONS
Environmentally Preferable