PYREX® Solution Bottles, Graduated, Corning
Supplier: Corning
1596-19L 1596-13L 1596-9L
16145-161CS 1101.92 CAD
16145-161 89091-150 89091-530
PYREX® Solution Bottles, Graduated, Corning
Bottles Solution Bottles
PYREX® borosilicate glass.
  • With double scale (liters and gallons) black enamel graduations
  • Graduated in 500 ml increments
  • Will withstand hot air (dry) or steam (wet) sterilization
  • Necks are tooled to increase mechanical strength and for uniform stopper fit
Solution bottles are designed for storage of solutions and are ideal for media preparation.