Eisco LDPE Wash Bottles with Sturdy Delivery Tube
Supplier: Wards
CH0181A CH0181D CH0181B CH0181C
470343-648EA 11.75 CAD
470343-648 470344-724 470344-768 470345-092
Eisco LDPE Wash Bottles with Sturdy Delivery Tube
Bottles Wash Bottles
Leak-proof dispensing of liquids.
  • Wash bottles made of flexible, premium quality low density polyethylene plastic (LDPE)
  • Translucent and unbreakable
  • High chemical durability and moderate heat tolerance
  • Can withstand temperatures between −50 to +80 °C
  • Good resistance to acids, alcohols and bases
  • Features a sturdy delivery tube that is integrated into the top screw cap
Features a sturdy delivery tube that is integrated into the top screw cap for leak-proof dispensing. The delivery tube is attached to an inner tube, which reaches the bottom of the bottle.