Eisco LDPE Wash Bottles with Flexible Delivery Tube
Supplier: Wards
CH0180C CH0180A CH0180D CH0180B
470344-572EA 7.45 CAD
470344-572 470344-902 470345-542 470345-768
Eisco LDPE Wash Bottles with Flexible Delivery Tube
Bottles Wash Bottles
Extra long, flexible delivery tube for better reach and aim when dispensing liquids.
  • Made of flexible, premium quality, low density polyethylene plastic (LDPE)
  • Translucent and unbreakable
  • High chemical durability and moderate heat tolerance
  • Can withstand temperatures between −50 to +80 °C
  • Good resistance to acids, alcohols and bases
  • Flexible delivery tube integrated into the top screw cap
Features a flexible delivery tube that is integrated into the top screw cap, allowing for better reach and aim when dispensing liquids. The tip of the tube has a removable cap for changing the stream. The delivery tube is attached to an inner tube, which reaches the bottom of the bottle.