VeriSpec® Certified Reference Material, KCl Conductivity Standards
Supplier: Ricca Chemical
RV588510-500A
RV588700-500A
RV588801-500A
RV588810-500A
RV588930-500A
RV588650-500A
RV588730-500A
RV588747-500A
RV588814-500A
RV588900-500A
RV588684-500A
RV588750-500A
RV588775-500A
RV588843-500A
RV588860-500A
RCRV588510500AEA
101.12
CAD
RCRV588510500A
RCRV588700500A
RCRV588801500A
RCRV588810500A
RCRV588930500A
RCRV588650500A
RCRV588730500A
RCRV588747500A
RCRV588814500A
RCRV588900500A
RCRV588684500A
RCRV588750500A
RCRV588775500A
RCRV588843500A
RCRV588860500A
VeriSpec® Certified Reference Material, KCl Conductivity Standards
Standards
Conductivity Standards
- Colorless Liquid
- Conductivity at 25 °C: See Ordering Table