Maleic acid ≥98%, Calbiochem®, Millipore®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

MX0100-20 MX0100-20 8.00380.9050
CA8.00380.9050EA 3175.04 CAD
TXMX0100-20WRG CA8.00380.9050 EM-MX0100-20
Maleic acid ≥98%, Calbiochem®, Millipore®
Maleic acid
Formula: C₄H₄O₄
MW: 116.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00063177
CAS Number: 110-16-7

Order Now

Specification Test Results

Assay (acidimetric) ≥ 99.0 %
Identity (IR) passes test
Melting range (lower value) ≥ 98 °C
Melting range (upper value) ≤ 103 °C
Spec. rotation α²0/D (c=5 in pyridine) -30.0 - -27.0 °

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR