Skolnik UN Certified Steel Tight Head Drums with 2" and ³/₄ " Fittings
Supplier: Qorpak
1749- KA605- 1727- 1657L- 1750- 1751L- 1422-
76579-204EA 355.9 CAD
76579-204 76579-184 76579-186 76579-176 76579-188 76579-200 76579-202
Skolnik UN Certified Steel Tight Head Drums with 2" and ³/₄ " Fittings
Barrels, Kegs, Drums
Qorpak® Skolnik UN certified steel drums are thicker, heavier, and stronger than industry standards require, offer extreme durability, and comply with domestic and international regulations.
  • Meets the rigorous UN packaging requirements of 49 CFR
  • Tested per the DOT and UN packaging test requirements
  • Heavier and stronger than industry standards
  • Better endures nail and forklift punctures than thinner drums
Do not risk place your volatile, expensive, irreplaceable, hazardous, or precious contents in any other vessel. Protect your products, reputation, and the environment with a superior quality drum.
Certifications: UN certified for liquids.