J.T.Baker® Cell Lysis Solution, Ultrapure Bioreagent
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
J.T.Baker® Cell Lysis Solution. Optimized multi-component, ready-to-use solution for extraction and recovery of AAV particles from HEK293 or SF9 cells. The product offers quantitative recovery through efficient cell lysis and protection of vector particles against shear stress. The solution is formulated with non-ionic detergents which are biodegradable and octyl phenol ethoxylate free.
- Comparable cell lysis performance to octyl phenol ethoxylate
- Improved protection of viral particles
- Reduced foaming and viscosity
- 0.2 µm filtered solution
Proprietary formulation. Not hazardous according to regulation 29 CFR 1910.1200 (OSHA HCS)
Chemical Specifications of Non-GMP Cell lysis solution:
Refractive Index: 1.1-1.5
Appearance: Passes Test
pH: For information only
Viscosity (25°C): For information only
Density: 0.9 – 1.2 g/mL
Total microbial count: No growth detected
UV-Vis (max Absorption) at 380-450 nm: For information only
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...