Klerwipe™ Sterile Non-Woven Dry Wipes
Supplier: Ecolab
Klerwipe™
53000342 53000345 53000343
76554-244CS 524.23 CAD
76554-244 76554-300 76554-246
Klerwipe™ Sterile Non-Woven Dry Wipes
Wipes Cleanroom Wipes Sterile Cleanroom Wipes
Ecolab Klerwipe™ Sterile Non-Woven Dry Wipes are low particulate wipes for cleanrooms. Made from a binder free, hydroentangled polyester/cellulose blend.
  • Gamma irradiated to a 10 -6 Sterility Assurance Level (SAL)
  • Header with Package Integrity testing
  • Do not contain chemical binders, leaving no undesirable residues
  • Highly absorbent, making them useful for applying cleaning solutions and to clean up spills
Klerwipe™ Sterile Non-Woven Dry Wipes are general purpose cleanroom wipe. Made from a binder free 55% cellulose/ 45% polyester hydroentangled nonwoven blend. This validated sterile version is ideal for cleaning applications during asepticprocessing activity. Excellent for use in Primary Engineering Controls (PEC) such as laminar air hoods, biological safety cabinets and compounding aseptic isolators where sterility of end product must be assured.
Suitable for use in controlled environments such as isolators and laminar flow cabinets.
Recommended for use in ISO Class 5 and above environments.
Certifications: Certificates of Sterility available.
Meets cGMP requirements for traceability.
Packaging: Double bagged.