Klerwipe™ Sterile 100% Polyester Dry Wipes
Supplier: Ecolab
Klerwipe™
53000350 53000351 53000347 53000348
76554-290CS 1572.67 CAD
76554-290 76554-292 76554-294 76554-296
Klerwipe™ Sterile 100% Polyester Dry Wipes
Wipes Cleanroom Wipes Sterile Cleanroom Wipes
Ecolab Klerwipe™ Sterile 100% Polyester Dry Wipes are heavy weight, knitted, ultrasonically sealed edge sterile wipes for cleanrooms.
  • Soft hand and edge for sensitive surfaces, abrasion resistant
  • Low particle and fiber generation, high level of cleanliness
  • Chemically compatible with disinfectants and cleaning solutions
  • Sealed edge, cleanroom laundered, knitted wipes offer higher purity and durability compared to nonwoven wipes
  • Packaging designed to minimize generation of particles
  • Highly absorbent, making them useful for applying cleaning solutions and to clean up spills
Klerwipe™ Sterile 100% Polyester Dry Wipes are general purpose cleanroom wipe. Ultrasonically sealed edge wipe is 100% knitted polyester and laundered and packaged in an ISO Class 4 clean-room.
Suitable for use in controlled environments such as isolators and laminar flow cabinets.
Recommended for use in ISO Class 3 and above environments.