Klercide™ Sterile Isolator Cleaning Tool Covers
Supplier: ECOLAB LIFE SCIENCES
92023004
76554-288CS 3892.73 CAD
76554-288
Klercide™ Sterile Isolator Cleaning Tool Covers
Mops
The isolator cleaning tool covers are made of 100% polyester.
  • Low particulate pads
  • Packaging is easy to open, even when wearing gloves
  • Supplied cleanroom processed, individually double bagged and sterilized by gamma irradiation