Klercide™ 70/30 IPA
Supplier: ECOLAB INC CA
6600101 6600136 6600097
CA76717-436CS 483.05 CAD
CA76717-436 CA76717-440 CA77322-218
Klercide™ 70/30 IPA
Cleaning Agents Disinfectants
Klercide™ 70/30 IPA is sterile 70% v/v Isopropyl alcohol blended with Water for Injection for daily disinfection of critical areas. Klercide™ 70/30 IPA is a one-step disinfectant IPA spray designed for disinfection of cleanroom environments, with trigger-spray format featuring the patented “bag-in-a-bottle” SteriShield™ Delivery System.
  • Efficacy - 0.2 micron-filtered 70% isopropyl alcohol solution blended with WFI (endotoxin level guaranteed to be <0.25 E.U./mL) to deliver Bactericidal, Fungicidal and Virucidal efficacy.
  • Daily Disinfection - 70/30 IPA
More Product Information