COSA™ CIP 96 Alkaline Cleaner
Supplier: ECOLAB LIFE SCIENCES
6600152 6600153
76554-206EA 673.52 CAD
76554-206 76554-208
COSA™ CIP 96 Alkaline Cleaner
Cleaning Agents Detergents and Surface Cleansers
COSA™ CIP 96 is a strong alkaline, surfactant-free cleaner.
  • Surfactant-free formula for ease of rinsing.
  • Chemical concentration can be dosed and monitored using conductivity.
  • With the addition of cleaning boosters in the COSA™ line, COSA™ CIP 96 is suitable for cleaning product lines with fatty and gel containing residues.
  • Supported by a complete validation package including techniques for determining acceptable residual levels and analytical methods for residue quantitation.
COSA™ CIP 96 is a strong alkaline, surfactant-free cleaner for cleaning in the pharmaceutical and personal care industries.