Spectrophotometers, Easy UV/Easy VIS, Mettler-Toledo
Supplier: Mettler Toledo
30705169 30705170
76580-222EA 10168.09 CAD
76580-222 76580-224
Spectrophotometers, Easy UV/Easy VIS, Mettler-Toledo
Spectrophotometers and Photometers UV/Visible Spectrophotometers
The Easy UV and Easy VIS spectrophotometers offer 3-in-1 functionality and is designed to greatly facilitate routine spectroscopic workflows.
  • Save space – The compact design similar to an A4 paper reduces clutter on your lab bench
  • Enjoy fast, compliant color measurement – Includes 30 color scales (e.g. CIELAB, APHA, Gardner, and more.)
  • Avoid errors – PathDetect™ securely checks the path length so your results are accurate
  • Test water quickly – The SQKitReader™ accessory unlocks over 150 Spectroquant® water test kit methods
  • Access methods easily – Built-in features such as a favorite list and LongClick™ speed up routine analyses
  • Clean up quickly thanks to a washable drip tray
  • The EasyPlus spectrophotometers are designed to reduce the effects of turbidity on measurements (please contact your local representative for culturing applications)
The instrument is simple and easy-to-use, and ensures accurate, repeatable measurements. Built-in methods also eliminate calculation and transcription errors.
Certifications: CE, CSA.