Evergreen Embedding Rings, Caplugs Evergreen
Supplier: Caplugs
258-4010-W10 258-4011-P10
76502-850CT 102.81 CAD
76502-850 76502-852
Evergreen Embedding Rings, Caplugs Evergreen
Embedding Cassettes Embedding Rings
Caplugs Evergreen embedding rings are for use in positioning histological tissues accurately in base molds. A treated sample is placed in the mold, the embedding ring is placed into position and the embedding medium is poured.
  • Rings are compatible with most commonly-used processing and storage systems
  • Etched labeling area is molded-in to ensure proper identification
  • Available in multiple colors
Precision-molded from premium-grade, chemically-inert, high-impact polystyrene for dimensional rigidity and sturdiness.