VWR® Cleanroom Gloves, Class 10, Latex, Non Sterile
Supplier: VWR International
76579-676 76579-678 76579-672 76579-674
76579-676CS 557.44 CAD
76579-676 76579-678 76579-672 76579-674
VWR® Cleanroom Gloves, Class 10, Latex, Non Sterile
Gloves
VWR® latex class 10 cleanroom non-sterile gloves are designed to provide exceptional protection and comfort in sterile controlled environments. These gloves meet stringent class 10 (ISO 4) standards, ensuring minimal particulate contamination. Ideal for pharmaceutical, biotech, and electronics applications, they offer enhanced dexterity, tactile sensitivity, and chemical resistance, making them a reliable choice for critical cleanroom tasks requiring precision and sterility.
  • Ambidextrous
  • Beaded cuff
  • Powder-free and silicone-free
  • Natural rubber latex
  • Low
More Product Information