Continuous Liner Kits, 23" Diameter, Rheo Flexibles (Division of Rheo Engineering)
Supplier: RHEO ENGINEERING LLC
4000684 4000993 40001050 4001441 4000098
76557-582BX 1109.38 CAD
76557-582 76557-640 76557-590 76557-596 76557-614
Continuous Liner Kits, 23" Diameter, Rheo Flexibles (Division of Rheo Engineering)
Mailers and Shippers
The Rheo Continuous Liner is an excellent pack-off solution for transferring materials from a work process into a transport container while maintaining a high level of containment. Continuous Liners come in various sizes and can be configured to work with existing process equipment.
  • Constructed from RheoFlex 302 polyethylene material
  • FDA approved for packaging
  • USP and EP compliant
  • 21 CFR 177.1520 compliant
  • Matte finish
  • Anti-static film
Continuous Liners keep the powder pack-off process clean and reduce the risk of cross contamination within the process suite. This technology can also provide a safe bag-in/bag-out option for transferring items into and out of an isolator.
Certifications: USP <661.1>, USP <661.2>, USP <88>, USP <87>, USP Class VI, EP 3.1.3, EU 10/2011, 94/62/EC, CONEG.
Ordering information: Store in box, out of UV light, in a cool and dry location.
Delivery information: Continuous Liner Kit comes with a Continuous Liner Pack with Support Ring, Stainless Steel Canister, Tri-Clamp with Support Flanges, and GripLoc Clamp. The Continuous Liner Pack is the replacement liner.