Transfer Isolators, 4 Gloves, Rheo Flexibles (Division of Rheo Engineering)
Supplier: RHEO ENGINEERING LLC
4001794 4001795 4001796 4002137 4002138 4002139
76557-606EA 5599.73 CAD
76557-606 76557-754 76557-578 76557-576 76557-550 76557-664
Transfer Isolators, 4 Gloves, Rheo Flexibles (Division of Rheo Engineering)
Glove Boxes and Glove Bags
The Rheo transfer isolator is a single use isolator that provides high containment operator protection during the handling of potentially hazardous powders. This quick deployment solution can be installed quickly and can be disposed of when finished eliminating the cleaning costs associated with traditional stainless steel isolators.
  • Constructed from RheoFlex 200 series film polyurethane material
  • FDA compliant
  • 21 CFR 177.1680 compliant
  • Clear and matte finish
  • Anti-static film
  • BSE/TSE compliant
The bungee strap system and the general flexibility of soft-wall enclosures provide ease of use for operators as they safely work with items inside of these safety isolators.
Caution: Store in box, out of UV light, in a cool, dry location.